Learn More
Hach Company All Weather Refrig Sampler
Catalog No. 5000208084 Shop All Hach Company Products
Encompass
AS950 All Weather Sampler with Heater Bundle-115V with 4 - 10 L BottlesIncludes1 ea AS950 controller on 115V All Weather base with controller compartment heater1 ea Bottle kit (AW040030) includes 4- 2 5 gallon poly bottles w/caps (2315) and distributor arm (8843)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)
Catalog No. 50-002-08084 Supplier Hach Company Supplier No. ASA.CXXX2X31XX
Add to Cart
Add to Cart
Request a Quote
AS950 All Weather Sampler with Heater Bundle-115V with 4 - 10 L BottlesIncludes1 ea AS950 controller on 115V All Weather base with controller compartment heater1 ea Bottle kit (AW040030) includes 4- 2 5 gallon poly bottles w/caps (2315) and distributor arm (8843)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)