Learn More
Hach Company All Weather Refrig Sampler
Catalog No. 5000207715 Shop All Hach Company Products
Encompass
AS950 All Weather Refrigerated Sampler-2 Sensor Port Options-115 V-Discrete (24x) 1 L PE Bottle Kit-digital pH SensorIncludes1 ea AS950 controller with sensor ports on 115V All Weather base1 ea Bottle kit (AW241000) includes 24- 1L poly bottles w/caps (737)-bottle tray (1511)-retainer (1322)-and distributor arm (8841)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)1 ea digital pH sensor (DPD2P1)1 ea adapter cable (9501200)
Catalog No. 50-002-07715 Supplier Hach Company Supplier No. ASA.CSXX1X413X
Add to Cart
Add to Cart
Request a Quote
AS950 All Weather Refrigerated Sampler-2 Sensor Port Options-115 V-Discrete (24x) 1 L PE Bottle Kit-digital pH SensorIncludes1 ea AS950 controller with sensor ports on 115V All Weather base1 ea Bottle kit (AW241000) includes 24- 1L poly bottles w/caps (737)-bottle tray (1511)-retainer (1322)-and distributor arm (8841)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)1 ea digital pH sensor (DPD2P1)1 ea adapter cable (9501200)