Learn More
Hach Company All Weather Refrig Sampler
Catalog No. 5000207738 Shop All Hach Company Products
Encompass
AS950 All Weather Refrigerated Sampler-2 Sensor Port Options-115 V-Heater-Composite 20 L PE Bottle-Submerged Area Velocity SensorIncludes1 ea AS950 controller with sensor ports on 115V All Weather base1 ea Bottle kit (AW010060) includes 1- 5 5 gallon poly bottle w/cap (6494)-composite tube support (8838) and full bottle shut-off (8847)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)1 ea oil-filled sub AV sensor (77064-030)1 ea AV9000S Analyzer (9504600)
Catalog No. 50-002-07738 Supplier Hach Company Supplier No. ASA.CSXX2X112X
Add to Cart
Add to Cart
Request a Quote
AS950 All Weather Refrigerated Sampler-2 Sensor Port Options-115 V-Heater-Composite 20 L PE Bottle-Submerged Area Velocity SensorIncludes1 ea AS950 controller with sensor ports on 115V All Weather base1 ea Bottle kit (AW010060) includes 1- 5 5 gallon poly bottle w/cap (6494)-composite tube support (8838) and full bottle shut-off (8847)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)1 ea oil-filled sub AV sensor (77064-030)1 ea AV9000S Analyzer (9504600)