Learn More
Medchemexpress LLC (all-Z)-6,9,12,15,18-heneicosapentaenoic acid ethyl ester | 131775-86-5 | 94.0% | 344.5 g/mol | C23H36O2 | 5MG
SDP
Catalog No. 5000238420
Encompass_Preferred
(all-Z)-6,9,12,15,18-Heneicosapentaenoic acid ethyl ester is the ethyl ester derivative of heneicosapentaenoic acid, an ω-3 polyunsaturated fatty acid. It is used as a research reagent and analytical reference standard in biochemical and lipid metabolism studies. Key identifiers include CAS 131775-86-5, formula C23H36O2, and molecular weight ≈344.5 g/mol.
  • Ethyl ester form for increased lipophilicity and stability.
  • Used as a reference standard in lipid metabolism and analytical assays.
  • Reported purity range 94-98%; verify certificate of analysis.
  • Typically supplied in small quantities suitable for analytical use (for example, 5 mg).
  • Characterized by CAS and standard structural identifiers for unambiguous identification.
Catalog No. 50-002-38420 Supplier Medchemexpress LLC Supplier No. HY1241185MG
(all-Z)-6,9,12,15,18-Heneicosapentaenoic acid ethyl ester is the ethyl ester derivative of heneicosapentaenoic acid, an ω-3 polyunsaturated fatty acid. It is used as a research reagent and analytical reference standard in biochemical and lipid metabolism studies. Key identifiers include CAS 131775-86-5, formula C23H36O2, and molecular weight ≈344.5 g/mol.
  • Ethyl ester form for increased lipophilicity and stability.
  • Used as a reference standard in lipid metabolism and analytical assays.
  • Reported purity range 94-98%; verify certificate of analysis.
  • Typically supplied in small quantities suitable for analytical use (for example, 5 mg).
  • Characterized by CAS and standard structural identifiers for unambiguous identification.