Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (all-Z)-8,11,14,17-eicosatetraenoic acid | 24880-40-8 | MFCD00216033 | >98.0% | 304.5 g/mol | C20H32O2 | 1MG
SDP

Catalog No. 5000238186
Encompass_Preferred

ω-3 arachidonic acid (all-Z-8,11,14,17-eicosatetraenoic acid) is a 20-carbon omega-3 polyunsaturated fatty acid supplied as a research reagent for lipid biology and enzymology. It has been reported to inhibit arachidonoyl-CoA synthetase and is provided in small milligram quantities suitable for analytical and enzymatic studies.

  • Molecular formula C20H32O2.
  • Molecular weight 304.5 g/mol.
  • Purity >98.0%.
  • Reported activity: inhibits arachidonoyl-CoA synthetase.
  • Intended use: lipid biology and enzymology research.
  • Supplied in milligram research quantities for analytical work.

Catalog No. 50-002-38186 Supplier Medchemexpress LLC Supplier No. HY1251401MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ω-3 arachidonic acid (all-Z-8,11,14,17-eicosatetraenoic acid) is a 20-carbon omega-3 polyunsaturated fatty acid supplied as a research reagent for lipid biology and enzymology. It has been reported to inhibit arachidonoyl-CoA synthetase and is provided in small milligram quantities suitable for analytical and enzymatic studies.

  • Molecular formula C20H32O2.
  • Molecular weight 304.5 g/mol.
  • Purity >98.0%.
  • Reported activity: inhibits arachidonoyl-CoA synthetase.
  • Intended use: lipid biology and enzymology research.
  • Supplied in milligram research quantities for analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.