Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Allantoic acid | 99-16-1 | ≥98.0% | 5 MG
SDP

Catalog No. 5000412107
Encompass_Preferred

Allantoic acid is an endogenous metabolite and a degradative product of uric acid, playing a role in purine metabolism. It has a molecular weight of 176.13 and a chemical formula of C4H8N4O4. It appears as a solid, typically white to off-white, and originates from human gut microbiota. This product is suitable for research use only, as it has not been fully validated for medical applications.

  • Degradative product of uric acid
  • Associated with purine metabolism
  • Solid, white to off-white appearance
  • Molecular weight of 176.13
  • Chemical formula C4H8N4O4
  • Initial source from human gut microbiota

Catalog No. 50-004-12107 Supplier Medchemexpress LLC Supplier No. HYB1514R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allantoic acid is an endogenous metabolite and a degradative product of uric acid, playing a role in purine metabolism. It has a molecular weight of 176.13 and a chemical formula of C4H8N4O4. It appears as a solid, typically white to off-white, and originates from human gut microbiota. This product is suitable for research use only, as it has not been fully validated for medical applications.

  • Degradative product of uric acid
  • Associated with purine metabolism
  • Solid, white to off-white appearance
  • Molecular weight of 176.13
  • Chemical formula C4H8N4O4
  • Initial source from human gut microbiota

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.