Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allantoin

Catalog No. 05670100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-670-100G 100 g
05-670-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-670-100G Supplier Sigma Aldrich Supplier No. 05670100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Allantoin

Specifications

CAS 97-59-6
Melting Point 230°C (lit.)
Molecular Formula C4H6N4O3
Linear Formula C4H6N4O3
MDL Number MFCD00005260
Quantity 100 g
Synonym NSC 7606; 5-Ureidohydantoin; Glyoxylic(acid) diureide
Molecular Weight (g/mol) 158.12
Percent Purity ≥98.0% (N)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.