Learn More
Selleck Chemical LLC Allantoin S3856-25mg
SDP
Catalog No. 503109016
Encompass_Preferred
Allantoin (Glyoxyldiureide 5-Ureidohydantoin) produced from uric acid is a major metabolic intermediate in most organisms including animals plants and bacteria
Catalog No. 50-310-9016 Supplier Selleck Chemical LLC Supplier No. S385625MG
Allantoin (Glyoxyldiureide 5-Ureidohydantoin) produced from uric acid is a major metabolic intermediate in most organisms including animals plants and bacteria