Learn More
Medchemexpress LLC Allantoin (5-Ureidohydantoin) | 97-59-6 | 98.9% | 158.12 | 100 MG
SDP
Catalog No. 5000365002
Encompass_Preferred
Allantoin is a skin conditioning agent that promotes healthy skin and stimulates new and healthy tissue growth. It possesses anti-oxidative and anti-inflammatory activities and can ameliorate chemically-induced pancreatic β-cell damage.
  • Promotes healthy skin and stimulates new tissue growth
  • Exhibits anti-oxidative and anti-inflammatory activities
  • Attenuates apoptosis and cytotoxicity, increasing β-cell viability
  • Activates imidazoline 3 (I3) receptors
  • Increases neuronal cell proliferation in the hippocampal dentate gyrus region
  • Lowers plasma glucose and increases plasma insulin levels
  • Decreases blood pressure
Catalog No. 50-003-65002 Supplier Medchemexpress LLC Supplier No. 100MGHYN0543
Allantoin is a skin conditioning agent that promotes healthy skin and stimulates new and healthy tissue growth. It possesses anti-oxidative and anti-inflammatory activities and can ameliorate chemically-induced pancreatic β-cell damage.
  • Promotes healthy skin and stimulates new tissue growth
  • Exhibits anti-oxidative and anti-inflammatory activities
  • Attenuates apoptosis and cytotoxicity, increasing β-cell viability
  • Activates imidazoline 3 (I3) receptors
  • Increases neuronal cell proliferation in the hippocampal dentate gyrus region
  • Lowers plasma glucose and increases plasma insulin levels
  • Decreases blood pressure