Learn More
Medchemexpress LLC Allantoin (5-ureidohydantoin) | 97-59-6 | MFCD00005260 | 158.12 | 25 G
SDP
Catalog No. 5000389471
Encompass_Preferred
Allantoin is a skin conditioning agent that promotes healthy skin and stimulates new and healthy tissue growth. It has reported anti-oxidative and anti-inflammatory activities. This product is for research use only.
  • Attenuates apoptosis and cytotoxicity
  • Increases the viability of STZ-induced β-cells
  • Decreases caspase-3 levels
  • Activates imidazoline 3 (I3) receptors
  • Increases latency time in passive avoidance tasks
  • Increases expression levels of phosphorylated PI3K, Akt, and GSK-3β
  • Increases neuronal cell proliferation in the hippocampal dentate gyrus
  • Lowers plasma glucose and increases plasma insulin levels in STZ-treated rats
  • Decreases blood pressure, inhibits cardiac contractility, heart rate, and increases peripheral blood flow in SHRs
Catalog No. 50-003-89471 Supplier Medchemexpress LLC Supplier No. 25GHYN0543
Allantoin is a skin conditioning agent that promotes healthy skin and stimulates new and healthy tissue growth. It has reported anti-oxidative and anti-inflammatory activities. This product is for research use only.
  • Attenuates apoptosis and cytotoxicity
  • Increases the viability of STZ-induced β-cells
  • Decreases caspase-3 levels
  • Activates imidazoline 3 (I3) receptors
  • Increases latency time in passive avoidance tasks
  • Increases expression levels of phosphorylated PI3K, Akt, and GSK-3β
  • Increases neuronal cell proliferation in the hippocampal dentate gyrus
  • Lowers plasma glucose and increases plasma insulin levels in STZ-treated rats
  • Decreases blood pressure, inhibits cardiac contractility, heart rate, and increases peripheral blood flow in SHRs