Learn More
Apexbio Technology LLC Allantoin 97-59-6 100mg
SDP
Catalog No. 5000096516
Encompass_Preferred
Allantoin (CAS 97-59-6) is a naturally occurring diureide of glyoxylic acid commonly isolated from the dried tubers of Dioscorea opposite Thunb This small molecule is known for its role in facilitating cell proliferation and promoting tissue repair attributed to its ability to modulate cellular response to injury and oxidative stress Allantoin is widely utilized in biomedical research for investigating mechanisms of wound healing anti-inflammatory responses and skin barrier restoration Proper storage in a sealed cool and dry environment is recommended to maintain compound stability
Catalog No. 50-000-96516 Supplier Apexbio Technology LLC Supplier No. N2301100
Allantoin (CAS 97-59-6) is a naturally occurring diureide of glyoxylic acid commonly isolated from the dried tubers of Dioscorea opposite Thunb This small molecule is known for its role in facilitating cell proliferation and promoting tissue repair attributed to its ability to modulate cellular response to injury and oxidative stress Allantoin is widely utilized in biomedical research for investigating mechanisms of wound healing anti-inflammatory responses and skin barrier restoration Proper storage in a sealed cool and dry environment is recommended to maintain compound stability