Learn More
Medchemexpress LLC Allantoin | 97-59-6 | MFCD00005260 | 158.12 | 5 G
SDP
Catalog No. 5000388804
Encompass_Preferred
Allantoin is a widely recognized skin conditioning agent that supports healthy skin and stimulates new tissue growth. It exhibits anti-oxidative and anti-inflammatory properties, and has been observed to attenuate apoptosis and cytotoxicity, enhancing the viability of STZ-induced β-cells in a dose-dependent manner. Additionally, allantoin decreases caspase-3 levels, boosts phosphorylated B-cell lymphoma 2 (Bcl-2) expression, and activates imidazoline 3 (I3) receptors.
  • Skin conditioning agent
  • Promotes healthy skin and new tissue growth
  • Exhibits anti-oxidative and anti-inflammatory activities
  • Attenuates apoptosis and cytotoxicity
  • Increases viability of STZ-induced β-cells
  • Activates imidazoline 3 (I3) receptors
Catalog No. 50-003-88804 Supplier Medchemexpress LLC Supplier No. HYN05435G
Add to Cart
Add to Cart
Allantoin is a widely recognized skin conditioning agent that supports healthy skin and stimulates new tissue growth. It exhibits anti-oxidative and anti-inflammatory properties, and has been observed to attenuate apoptosis and cytotoxicity, enhancing the viability of STZ-induced β-cells in a dose-dependent manner. Additionally, allantoin decreases caspase-3 levels, boosts phosphorylated B-cell lymphoma 2 (Bcl-2) expression, and activates imidazoline 3 (I3) receptors.
  • Skin conditioning agent
  • Promotes healthy skin and new tissue growth
  • Exhibits anti-oxidative and anti-inflammatory activities
  • Attenuates apoptosis and cytotoxicity
  • Increases viability of STZ-induced β-cells
  • Activates imidazoline 3 (I3) receptors