Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allantoin, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015996 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5996 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5996 Supplier MilliporeSigma Supelco Supplier No. 9379150MG
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 97-59-6
Physical Form Neat
Linear Formula C4H6N4O3
MDL Number MFCD00005260
Melting Point 230°C
Synonym 5-Ureidohydantoin; Glyoxylic(acid) diureide
Solubility Soluble in water
Type Analytical Standard
Quantity 50 mg
Formula Weight 158.12
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.