Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allantoin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015995 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5995 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5995 Supplier MilliporeSigma Supelco Supplier No. PHR1364500MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 97-59-6
Linear Formula C4H6N4O3
MDL Number MFCD00005260
Melting Point 230°C
Synonym Allantoin; 5-Ureidohydantoin; Glyoxylic(acid) diureide
Type Certified Reference Material
Quantity 500 mg
Formula Weight 158.12
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.