Learn More
CPC Scientific H-Gly-Phe-Lys-Asn-Val-Glu-Met-Met-Thr-Ala-Arg-Gly-Phe-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific INSE002B
Encompass
The neurohormone allatotropin, GFKNVEMMTARGF-amide, has been isolated from head extracts of the adult tobacco hornworm, Manduca sexta. It strongly stimulates the release of juvenile hormone from the corpora allata in vitro.ONE-LETTER SEQUENCE: GFKNVEMMTARGFMOLECULAR FORMULA: C65H103N19O17S2MOLECULAR WEIGHT:1486.78STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [120928-88-3], RESEARCH AREA: NeuropeptidesREFERENCES: H.Kataoka et al., Science, 243, 1481 (1989); P.E.A.Teal, Peptides, 23, 663 (2002)
Catalog No. 50-210-9604