Learn More
Aviva Systems Biology Corp ALLC Antibody - N-terminal region (ARP57249_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57249_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALLC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ALLC. Alternate names: ALCApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit
Catalog No. 50-014-62112