Learn More
Cayman Chemical allcis4 7 10 13 16Docosapenta
SDP
Catalog No. 502888958
Encompass_Preferred
An ω-6 isomer of docosapentaenoic acid formed by the elongation and desaturation of arachidonic acid; diminished during fatty acid desaturase syndrome and increased in non-alcoholic steatohepatitis
Catalog No. 50-288-8958 Supplier Cayman Chemical Supplier No. 1000833510 MG
An ω-6 isomer of docosapentaenoic acid formed by the elongation and desaturation of arachidonic acid; diminished during fatty acid desaturase syndrome and increased in non-alcoholic steatohepatitis