Learn More
Cayman Chemical allcis4 7 10 13 16Docosapenta
SDP
Supplier:  Cayman Chemical 1000833525 MG
Encompass_Preferred
An ω-6 isomer of docosapentaenoic acid formed by the elongation and desaturation of arachidonic acid; diminished during fatty acid desaturase syndrome and increased in non-alcoholic steatohepatitis
Catalog No. 50-288-7364