Learn More
Andwin Scientific ALLEGIANCE TUBING
SDP
Catalog No. NC9755498
Encompass_Preferred
The MediVac Suction Connecting Tubes measure 6 feet in length and 3/16 Inch (5mm) for its inner diameter. This product is able to withstand collapse up to 22 inch Hg vacuum. It is also sterile and non-conductive.
Catalog No. NC9755498 Supplier Andwin Scientific Supplier No. N56A
The MediVac Suction Connecting Tubes measure 6 feet in length and 3/16 Inch (5mm) for its inner diameter. This product is able to withstand collapse up to 22 inch Hg vacuum. It is also sterile and non-conductive.