Learn More
Beckman Coulter Beckman Coulter Allegra X-30 Clinical Benchtop Centrifuge, 1.6 L (MPN B06314)
Catalog No. NC1827577
Encompass
The Beckman Coulter Allegra X-30 Clinical Centrifuge (MPN B06314) is a compact, high-performance benchtop centrifuge designed for clinical and general laboratory sample preparation, including in vitro diagnostic workflows. It delivers high speeds and RCF for efficient separations and supports a broad rotor library for multiple sample formats.
  • Compact benchtop design saves laboratory space.
  • High-speed performance up to 16,000-18,000 rpm for rapid separation.
  • Maximum RCF up to 29,756 x g for efficient pellet formation.
  • 1.6 L maximum capacity accommodates various sample sizes.
  • Supports fixed-angle and swinging-bucket rotors for versatile applications.
  • Operates on 100 V, 50-60 Hz power suitable for clinical environments.
  • Simple setup and intuitive controls reduce run time and training needs.
This item is not returnable. View return policy
Catalog No. NC1827577 Supplier Beckman Coulter Supplier No. B06314
Add to Cart
Add to Cart
Request a Quote
The Beckman Coulter Allegra X-30 Clinical Centrifuge (MPN B06314) is a compact, high-performance benchtop centrifuge designed for clinical and general laboratory sample preparation, including in vitro diagnostic workflows. It delivers high speeds and RCF for efficient separations and supports a broad rotor library for multiple sample formats.
  • Compact benchtop design saves laboratory space.
  • High-speed performance up to 16,000-18,000 rpm for rapid separation.
  • Maximum RCF up to 29,756 x g for efficient pellet formation.
  • 1.6 L maximum capacity accommodates various sample sizes.
  • Supports fixed-angle and swinging-bucket rotors for versatile applications.
  • Operates on 100 V, 50-60 Hz power suitable for clinical environments.
  • Simple setup and intuitive controls reduce run time and training needs.