Learn More
Beckman Coulter Beckman Coulter Allegra X-30 High Cap Pkg
Catalog No. 501631959
Encompass_Preferred
High Capacity Cell Culture Application Package: Allegra X-30, IVD, 120 V, 60 Hz, Ambient Benchtop Centrifuge , 4x 400 mL SX4400 Swinging-bucket Rotor,15 mL Conical Adapters (2 sets of 2), 50 mL Conical Adapters (2 sets of 2). Max. tube capacity: 15 mL = 36 tubes / 50 mL = 16 tubes (Not compatible with optional BioCertified bucket covers.)
This item is not returnable. View return policy
Catalog No. 50-163-1959 Supplier Beckman Coulter Supplier No. B08705
Add to Cart
Add to Cart
Request a Quote
High Capacity Cell Culture Application Package: Allegra X-30, IVD, 120 V, 60 Hz, Ambient Benchtop Centrifuge , 4x 400 mL SX4400 Swinging-bucket Rotor,15 mL Conical Adapters (2 sets of 2), 50 mL Conical Adapters (2 sets of 2). Max. tube capacity: 15 mL = 36 tubes / 50 mL = 16 tubes (Not compatible with optional BioCertified bucket covers.)