Learn More
Beckman Coulter Beckman Coulter Allegra X-30R w F2402H
Catalog No. 501631956
Encompass_Preferred
Molecular Performance Application Package: Allegra X-30R, IVD, 120 V, 60 Hz, Refrigerated Benchtop Centrifuge with F2402H 24 x 1.5/2.0 mL BioCertified Fixed-angle Rotor
This item is not returnable. View return policy
Catalog No. 50-163-1956 Supplier Beckman Coulter Supplier No. B08547
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Molecular Performance Application Package: Allegra X-30R, IVD, 120 V, 60 Hz, Refrigerated Benchtop Centrifuge with F2402H 24 x 1.5/2.0 mL BioCertified Fixed-angle Rotor