Learn More
Beckman Coulter Beckman Coulter Allegra X-5 Clinical Package
Catalog No. 501631955
Encompass_Preferred
Allegra X-5 Clinical Sample Prep Package: Allegra X-5 ventilated benchtop centrifuge, IVD, 120 V, 60 Hz, SX4700 Swinging Bucket Rotor, Adapters for 13mm Tubes (Set of 4), Adapters for 17mm Tubes (Set of 4)
This item is not returnable. View return policy
Catalog No. 50-163-1955 Supplier Beckman Coulter Supplier No. B30589
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Allegra X-5 Clinical Sample Prep Package: Allegra X-5 ventilated benchtop centrifuge, IVD, 120 V, 60 Hz, SX4700 Swinging Bucket Rotor, Adapters for 13mm Tubes (Set of 4), Adapters for 17mm Tubes (Set of 4)