Learn More
Allegro™ Banana Oil Fit-Test Ampules
Catalog No. 19060428
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Used for testing respirator fit
Catalog No. For Use With (Application)
19-060428 Used for testing respirator fit
1 options
Catalog No. 19-060428 Supplier Allegro™ Supplier No. 0201
Banana oil (isoamyl acetate) has a strong, recognizable odor to quickly indicate the lack of proper respirator fit.
  • Banana oil (isoamyl acetate) is the substance originally used for testing respirator fit; its strong, recognizable odor quickly indicates the lack of proper fit
  • Available in two fit test kits and Fit Check Ampules that provide a fast and easy method of on-the-spot qualitative fit testing
Order Info
The Standard Kit comes in a handy carry bag and includes two 1 oz. (30mL) bottles of testing agent, a vinyl test enclosure, measuring pipette, and paper towels. The Deluxe Kit comes in a convenient storage case and contains everything necessary to perform a proper fit test, including an 8 1oz. (30mL) bottles of testing solution, a fit test tent, jars, paper towels, disposable bags, and other necessary components. Replacement banana oil solution is available in 1 oz. (30mL) bottles.
Specifications
Product Type Banana Oil Ampule
For Use With (Application) Used for testing respirator fit
Model 0201
Quantity 10/Pk.