Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allegro™ Bitrex Fit Test Kit: Solution

Catalog No. p-3202652
Change view
Click to view available options
Product Type:
Sensitivity Solution
Test Solution
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-088-509 Sensitivity Solution
19-088-510 Test Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-088-509 Supplier Allegro™ Supplier No. 204111K
Only null left
Add to Cart
Add to Cart

Offers qualitative OSHA compliant fit testing for disposable and reusable dust and mist respirators.

Bitrex Sensitivity Solution and Bitrex Test Solution

  • Box of 6 glass ampules each
  • With pre-drilled plastic cap for easy pouring
  • Plastic protection from broken glass
  • One-time use
  • Averages 3 to 5 fit tests per tube
  • Contains 2.5cc of solution

Specifications

Product Type Sensitivity Solution
Certifications/Compliance OSHA
For Use With (Equipment) Allegro Bitrex Fit Test Kit (2041)
Model 2041-11K
Quantity 6/Pk.
Type (Solution) Sensitivity solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.