Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allegro™ Bitrex Fit Test Kit

Catalog No. 19088508
Change view
Click to view available options
Bitrex Fit Test Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-088-508 Bitrex Fit Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-088-508 Supplier Allegro™ Supplier No. 2041/DEL

Offers qualitative OSHA compliant fit testing for disposable and reusable dust and mist respirators.

 

  • Bitrex creates bitter taste that indicates breakthrough on the respirator fit
  • New disposable glass ampules make pouring solution into nebulizers easier and reduce the chance of contamination
  • Additional Bitrex sensitivity solution ampules and Bitrex test solution ampules also available

 

Kit includes:

Test Hood

  • Poly-coated
  • Clear viewing window
  • Surged seams
  • Reusable
Sensitivity Nebulizer and Test Nebulizer
  • Natural Latex rubber air bulb for easy operation
  • Converts 5cc (mL) of liquid into aerosol 0.3 to 5.0 microns particle size
  • Complete with extra jet and replaceable plastic caps
  • Break-resistant polycarbonate plastic (Lexan™) that easily disassembles for washing
  • Dimensions: 2 x 2 x 7 in. (5 x 5 x 17.7cm)
Bitrex Sensitivity Solution and Bitrex Test Solution
  • Box of 6 glass ampules each
  • With pre-drilled plastic cap for easy pouring
  • Plastic protection from broken glass
  • One-time use
  • Averages 3 to 5 fit tests per tube
  • Contains 2.5cc of solution

 

Specifications

Product Type Bitrex Fit Test Kit
Certifications/Compliance OSHA
For Use With (Application) For disposable and reusable dust and mist respirators
Includes Test hood, 2 x 2 x 7 in. sensitivity nebulizer, test nebulizer, 6 ampules of sensitivity solution and 6 ampules of test solution
Model 2041
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.