Learn More
MSC Allegro Ducting Storage Bag, 8 in Ducting, Polyester, 1 ea
Catalog No. 5000793618
Encompass
This durable polyester storage bag is designed for the easy storage and transportation of ducting, providing a tightly controlled fit. It can accommodate up to 25 ft. of 8-in. ducting.
  • Heavy-duty polyester material
  • Reinforced carry handle
  • Shoulder strap
  • Dual zipper closure for easy access
  • Cylinder shape for tight fit
  • Black color fabric with white lettering
  • Dimensions: 25 in L x 14 in W x 14 in H
Catalog No. 50-007-93618 Supplier MSC Supplier No. 73511958
This durable polyester storage bag is designed for the easy storage and transportation of ducting, providing a tightly controlled fit. It can accommodate up to 25 ft. of 8-in. ducting.
  • Heavy-duty polyester material
  • Reinforced carry handle
  • Shoulder strap
  • Dual zipper closure for easy access
  • Cylinder shape for tight fit
  • Black color fabric with white lettering
  • Dimensions: 25 in L x 14 in W x 14 in H