Learn More
MSC Allegro Fit Test Kit, Bitter, Disposable/Reusable Respirator, 1 Kit
Catalog No. 5000792375
Encompass
This fit test kit is designed for quantitative respiratory fit testing, ensuring the proper seal of respirators. It uses a bitter solution to verify compatibility with various types of dust and mist respirators.
  • Includes sensitivity nebulizer
  • Includes test nebulizer
  • Includes test hood
  • Includes box of test solution
  • Includes box of sensitivity solution
  • Compatible with disposable and reusable dust and mist respirators
Catalog No. 50-007-92375 Supplier MSC Supplier No. 82707365
This fit test kit is designed for quantitative respiratory fit testing, ensuring the proper seal of respirators. It uses a bitter solution to verify compatibility with various types of dust and mist respirators.
  • Includes sensitivity nebulizer
  • Includes test nebulizer
  • Includes test hood
  • Includes box of test solution
  • Includes box of sensitivity solution
  • Compatible with disposable and reusable dust and mist respirators