Learn More
MSC Allegro Industries Carbon Monoxide Calibration Gas, 0.0001-0.0999 percent, 17 L
Catalog No. 5000782038
Encompass
This calibration span gas is designed for calibrating analytical instrumentation, specifically for carbon monoxide monitors. It comes in a 17 L cylinder and consists of a precise gas mixture.
  • Contains carbon monoxide (0.0001-0.0999%), oxygen (20.9%), and nitrogen (69.11-79.09%)
  • Colorless and odorless gas
  • Non-flammable mixture
  • Very slight solubility
  • Ideal for instrument calibration
  • Manufactured in the USA
Catalog No. 50-007-82038 Supplier MSC Supplier No. 02713204
This calibration span gas is designed for calibrating analytical instrumentation, specifically for carbon monoxide monitors. It comes in a 17 L cylinder and consists of a precise gas mixture.
  • Contains carbon monoxide (0.0001-0.0999%), oxygen (20.9%), and nitrogen (69.11-79.09%)
  • Colorless and odorless gas
  • Non-flammable mixture
  • Very slight solubility
  • Ideal for instrument calibration
  • Manufactured in the USA