Learn More
MSC Allegro Remote CO Alarm with Strobe, 10 PPM, 1 ea
Catalog No. 5000781378
Encompass
This remote CO alarm and strobe light system provides an additional warning mechanism for users of a filtration panel when carbon monoxide levels exceed 10 PPM. It connects to any filtration panel with an included 50 ft cord/cable, allowing the alarm to be positioned closer to working areas for timely alerts.
  • Loud 90 decibel audio alarm
  • Highly visible red strobe light
  • Dimensions: 9.5 in W x 10 in H x 8 in D
  • Includes 50 ft cord/cable
  • Made in the USA
Catalog No. 50-007-81378 Supplier MSC Supplier No. 45314077
This remote CO alarm and strobe light system provides an additional warning mechanism for users of a filtration panel when carbon monoxide levels exceed 10 PPM. It connects to any filtration panel with an included 50 ft cord/cable, allowing the alarm to be positioned closer to working areas for timely alerts.
  • Loud 90 decibel audio alarm
  • Highly visible red strobe light
  • Dimensions: 9.5 in W x 10 in H x 8 in D
  • Includes 50 ft cord/cable
  • Made in the USA