Learn More
Allegro™ Reusable Respirator Bag
Catalog No. 19041317
Encompass_Preferred
Change view
Click to view available options
Closure Type:
Zipper Closure
Catalog No. Closure Type
19-041317 Zipper Closure
1 options
Catalog No. 19-041317 Supplier Allegro™ Supplier No. 2000
Add to Cart
Add to Cart
Transparent 10 mil polypropylene reusable bag is durable and made for half or full facepiece respirators.
  • Bag is large enough to keep full-facepiece or half-mask respirators contaminant-free
  • Zipper assuring positive closure
  • Cleaning instructions are printed on the bag
Specifications
Product Type Reusable Respirator Storage Bag
Closure Type Zipper Closure
Color Clear
Model 2000