Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allegro™ SCBA Bracket for Standard Wall Case

Catalog No. 19029866
Encompass_Preferred
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-029866 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-029866 Supplier Allegro™ Supplier No. 411000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

White bracket for use with SCBA wall mount.

  • For SCBA wall-mount storage use
  • 45.72 x 17.02 x 0.25 cm

Specifications

Product Type SCBA Standard Bracket
Color White
Material Powder-coated Steel
Model 411000
For Use With (Equipment) Standard Wall Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.