Learn More
MSC Allegro Smoke Fit Test Kit, Compatible with Ampule, 1 Kit
Catalog No. 5000781926
Encompass
This Smoke Fit Test Kit is designed to perform qualitative fit testing for respirators using a smoke-generating ampule. It provides a reliable method for verifying proper respirator fit in various work environments.
  • Includes breaker tool, hand operated air bulb, poly tubing, and smoke tubes
  • Supplied with a vinyl protective pouch for storage and transport
  • Compatible with ampule-based test method
  • Suitable for qualitative respirator fit testing
Catalog No. 50-007-81926 Supplier MSC Supplier No. 55345003
This Smoke Fit Test Kit is designed to perform qualitative fit testing for respirators using a smoke-generating ampule. It provides a reliable method for verifying proper respirator fit in various work environments.
  • Includes breaker tool, hand operated air bulb, poly tubing, and smoke tubes
  • Supplied with a vinyl protective pouch for storage and transport
  • Compatible with ampule-based test method
  • Suitable for qualitative respirator fit testing