Learn More
Allegro™ Respirator Wipes
Catalog No. 19037680
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
15.2 x 19 cm (6 x 7.5 in.)
Catalog No. Dimensions (L x W)
19-037-680 15.2 x 19 cm (6 x 7.5 in.)
1 options
Catalog No. 19-037-680 Supplier Allegro™ Supplier No. 5001
Easy to use wipes for respirator and other personal safety equipment clean-up.
  • Benzalkonium chloride and alcohol reduce hazard of contamination
Specifications
Product Type Respirator Wipes
Model 5001
Solution Type Benzalkonium chloride
For Use With (Application) Personal safety equipment clean-up
Dimensions (L x W) 15.2 x 19 cm (6 x 7.5 in.)