Learn More
TA Instruments Allen Wrench. 050 Ball End, Precision Tool for PCA and DSC Accessories, Enables Secure Fastening and Adjustment of Instrument Components
Catalog No. 5000580722 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Wrench Allen .050 Ball End is designed for fine adjustments and secure fastening of PCA and DSC accessory components. Its ball end tip allows entry at various angles, improving accessibility in tight spaces during assembly and maintenance tasks. Engineered from durable materials, this precision tool provides long-term reliability and accuracy, ensuring instrument accessories remain properly aligned and secure. It is an essential support tool for laboratories managing sensitive photocalorimetry and calorimetry applications.
Catalog No. 50-005-80722 Supplier TA Instruments Supplier No. 265981.001
Waters TA Instruments Wrench Allen .050 Ball End is designed for fine adjustments and secure fastening of PCA and DSC accessory components. Its ball end tip allows entry at various angles, improving accessibility in tight spaces during assembly and maintenance tasks. Engineered from durable materials, this precision tool provides long-term reliability and accuracy, ensuring instrument accessories remain properly aligned and secure. It is an essential support tool for laboratories managing sensitive photocalorimetry and calorimetry applications.