Learn More
Waters Corp Waters Bottle Tray Kit, Larger Capacity, HPLC, Separations Modules
Catalog No. 50791668 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Alliance Bottle Tray Kit is a larger capacity solvent tray designed as an option for all Separations Modules. It works by relocating the detector directly above the column heater or column heater/cooler.
  • Minimizes ambient temperature effects on column to detector flow cell tubing.
  • Creates more space for larger solvent bottles.
  • Enhances routing of solvent and wash line tubings from the top of the system down into the module.
Catalog No. 50-791-668 Supplier Waters Corp Supplier No. 205000329
The Alliance Bottle Tray Kit is a larger capacity solvent tray designed as an option for all Separations Modules. It works by relocating the detector directly above the column heater or column heater/cooler.
  • Minimizes ambient temperature effects on column to detector flow cell tubing.
  • Creates more space for larger solvent bottles.
  • Enhances routing of solvent and wash line tubings from the top of the system down into the module.