Learn More
Waters Corp Waters Interactive System Support CD, For Alliance System, Maintenance Information
Catalog No. 50466693 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Alliance Interactive System Support CD provides essential overview and maintenance information, updated to ensure optimal performance and maximum uptime for your Alliance System. It is structured into four main areas for comprehensive support.
  • Alliance system section: introduces major assemblies, programmability, communications, and e2695 separations module options.
  • Optimizing performance section: covers solvent usage, major components, and performance maintenance procedures.
  • Maintenance section: offers instructional videos and PDFs for specific procedures on components like the e2695 separations module, 2998 photodiode array, 2414 refractive index, 2489 UV/visible, 2475 multi-wavelength fluorescence, and 2424 evaporative light scattering detector.
  • Service and support section: provides web links to additional support programs and resources.
Catalog No. 50-466-693 Supplier Waters Corp Supplier No. 715000116
This Alliance Interactive System Support CD provides essential overview and maintenance information, updated to ensure optimal performance and maximum uptime for your Alliance System. It is structured into four main areas for comprehensive support.
  • Alliance system section: introduces major assemblies, programmability, communications, and e2695 separations module options.
  • Optimizing performance section: covers solvent usage, major components, and performance maintenance procedures.
  • Maintenance section: offers instructional videos and PDFs for specific procedures on components like the e2695 separations module, 2998 photodiode array, 2414 refractive index, 2489 UV/visible, 2475 multi-wavelength fluorescence, and 2424 evaporative light scattering detector.
  • Service and support section: provides web links to additional support programs and resources.