Learn More
Enterprise Technology Solutions Alliance Rubber Company Rubber Bands, Size 33, 1/4 lb., 3-1/2"x1/8",Approx. 150/BX
SDP
Catalog No. 5000832292
Encompass_Preferred
This No. 33 rubber band for everyday use offers a mid-range rubber content. Advantage rubber bands are recommended for industrial applications where a sturdy band with higher tensile strength is required. The number count of the rubber bands is approximate.
Catalog No. 50-008-32292 Supplier Enterprise Technology Solutions Supplier No. ALL26339
This No. 33 rubber band for everyday use offers a mid-range rubber content. Advantage rubber bands are recommended for industrial applications where a sturdy band with higher tensile strength is required. The number count of the rubber bands is approximate.