Learn More
Waters Corp Waters Rebuild Kit, Alliance Solvent Manager, HPLC System, Annual Maintenance
Catalog No. 50792940 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit for the Alliance Solvent Manager is designed to ensure the optimal operation of Alliance HPLC Systems. This comprehensive kit provides all necessary components for annual preventive maintenance, helping to maintain peak instrument performance and data integrity.
  • Includes components for pump head, check valve, and and general rebuild.
  • Compatible with HPLC systems.
  • Designed for annual replacement.
  • Suitable for 2695 Separations Module, 2695 D Dissolution Separations Module, and 2795 Separations Module.
  • Contains diffuser, plunger seals, face seals, wash tube, plunger assembly, sealwash face seals, sealwash plunger seals, check valve cartridges, solvent filter, and a filter insert.
Catalog No. 50-792-940 Supplier Waters Corp Supplier No. 700004073
The Waters Performance Maintenance Kit for the Alliance Solvent Manager is designed to ensure the optimal operation of Alliance HPLC Systems. This comprehensive kit provides all necessary components for annual preventive maintenance, helping to maintain peak instrument performance and data integrity.
  • Includes components for pump head, check valve, and and general rebuild.
  • Compatible with HPLC systems.
  • Designed for annual replacement.
  • Suitable for 2695 Separations Module, 2695 D Dissolution Separations Module, and 2795 Separations Module.
  • Contains diffuser, plunger seals, face seals, wash tube, plunger assembly, sealwash face seals, sealwash plunger seals, check valve cartridges, solvent filter, and a filter insert.