Learn More
Waters Corp Waters Corporation Total Recovery Vial, Clear Glass, 12 x 32 mm, Snap Neck, 1 mL
Catalog No. 50818628 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Clear Glass 12 x 32 mm Snap Neck Total Recovery Vial, with Cap and PTFE/silicone Septum, is engineered for use with the side drawport needle and factory needle draw depth settings of the Alliance 2690/2695 HPLC. It offers a maximum sample capacity of 1 mL with a minimum residual volume of about 9 μL.
  • Securely holds analytes, samples, and solutions.
  • Engineered for use with side drawport needle and factory needle draw depth settings of Alliance 2690/2695 HPLC.
  • Maximum sample capacity: 1 mL; Minimum residual volume: ~9 μL.
  • PTFE/Silicone septum provides excellent resealing, suitable for multiple injections and storage.
  • Operating temperature range: -40 °C to 200 °C.
  • Snap cap mechanism ensures excellent sealing without tools.
  • Includes vial, cap, and septum; 100 units per package.
Catalog No. 50-818-628 Supplier Waters Corp Supplier No. 186000234
This Clear Glass 12 x 32 mm Snap Neck Total Recovery Vial, with Cap and PTFE/silicone Septum, is engineered for use with the side drawport needle and factory needle draw depth settings of the Alliance 2690/2695 HPLC. It offers a maximum sample capacity of 1 mL with a minimum residual volume of about 9 μL.
  • Securely holds analytes, samples, and solutions.
  • Engineered for use with side drawport needle and factory needle draw depth settings of Alliance 2690/2695 HPLC.
  • Maximum sample capacity: 1 mL; Minimum residual volume: ~9 μL.
  • PTFE/Silicone septum provides excellent resealing, suitable for multiple injections and storage.
  • Operating temperature range: -40 °C to 200 °C.
  • Snap cap mechanism ensures excellent sealing without tools.
  • Includes vial, cap, and septum; 100 units per package.