Learn More
Lkt Laboratories CAS 539-86-6, ≥98%, This product is a solution of methanol:water:formic acid (60:40:0.1) and allicin at 10mg/mL. Organosulfur found in garlic, binds DNA;.
SDP
Catalog No. NC2525886
Encompass_Preferred
This product is a solution of methanol:water:formic acid (60:40:0.1) and allicin at 10mg/mL. Allicin is an thiocyanate found in garlic; it exhibits a wide variety of properties, including anticancer, antioxidative, antihypertensive, anti-arrhythmic, anti-parasitic, and anti-diabetic activities. In vitro, allicin binds cellular nucleic acid, primarily nitrogenous bases and phosphate backbones, and induces autophagy and apoptosis, leading to cell death. The cardiovascular activities of allicin stem from its ability to decrease systolic blood pressure and triglyceride levels and to shorten action potential duration through inhibition of L-type voltage-gated Ca2+ channels and activation of inward rectifying K+ channels in animal models.
Catalog No. NC2525886 Supplier Lkt Laboratories Supplier No. A4440
This product is a solution of methanol:water:formic acid (60:40:0.1) and allicin at 10mg/mL. Allicin is an thiocyanate found in garlic; it exhibits a wide variety of properties, including anticancer, antioxidative, antihypertensive, anti-arrhythmic, anti-parasitic, and anti-diabetic activities. In vitro, allicin binds cellular nucleic acid, primarily nitrogenous bases and phosphate backbones, and induces autophagy and apoptosis, leading to cell death. The cardiovascular activities of allicin stem from its ability to decrease systolic blood pressure and triglyceride levels and to shorten action potential duration through inhibition of L-type voltage-gated Ca2+ channels and activation of inward rectifying K+ channels in animal models.