Learn More
Lkt Laboratories Allicin, aqueous solution, ≥98%, 1MG
SDP
Supplier:  Lkt Laboratories A44411MG
Encompass_Preferred
Organosulfur found in garlic; Kir K+ channel activator, L-type Ca2+ channel blocker. CAS: 539-86-6. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC0771584