Learn More
Chemglass Life Sciences Medium Condenser Only, Extraction Apparatus, Soxhlet, 45/50
SDP
Catalog No. CG136805
Encompass_Preferred
Medium condenser. Part of the CG1368 series. Condenser has an enlarged uptake tube to improve condensing capacity.
Catalog No. CG136805 Supplier Chemglass Life Sciences Supplier No. CG136805
Medium condenser. Part of the CG1368 series. Condenser has an enlarged uptake tube to improve condensing capacity.