Learn More
Med Vet International Miltex Vantage Allis Tissue Forceps, 6in, with 4x5 Teeth, 4.5mm Wide Jaws
Supplier:  Med Vet International MILV9166
Encompass_Preferred
The Miltex Vantage V916-6 Allis tissue forceps are made from high quality Japanese stainless steel and measure 6" in length. They have a finger ring handle with a ratchet lock and straight jaws 4.5mm wide that feature 4 x 5 teeth.
Catalog No. 50-255-9475