Learn More
Sklar Instruments Allis Tissue Forceps, Sklar, 6 in., 3x4 Teeth, Straight, Stainless Steel. EA/1.
SDP
Catalog No. 501438263 Shop All Sklar Instruments Products
Encompass_Preferred
Allis Tissue Forceps, Sklar, 6 in., 3x4 Teeth, Straight, Stainless Steel. Sklar Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue during surgical procedures. They are commonly used with tonsil, vaginal, breast, and thyroid tissue. Available straight or angled, this instrument is offered in a variety of lengths. The tip end style varies to meet the needs of the procedure or preferences of the surgeon. This product is straight with 3x4 teeth and an overall length of 6 inches.
This item is not returnable. View return policy
Catalog No. 50-143-8263 Supplier Sklar Instruments Supplier No. 362060
Allis Tissue Forceps, Sklar, 6 in., 3x4 Teeth, Straight, Stainless Steel. Sklar Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue during surgical procedures. They are commonly used with tonsil, vaginal, breast, and thyroid tissue. Available straight or angled, this instrument is offered in a variety of lengths. The tip end style varies to meet the needs of the procedure or preferences of the surgeon. This product is straight with 3x4 teeth and an overall length of 6 inches.