Learn More
Medchemexpress LLC Allitol (Allodulcitol) | 488-44-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN28405MG
Encompass_Preferred
Allitol (Allodulcitol) is an orally active rare sugar alcohol discovered in *Itea virginica L* and *Tylopilus plumbeoviolaceus*. It exhibits anti-obesity and hypoglycemic properties, reducing carcass and total body fat mass, and increasing cecal weight and short-chain fatty acid production. It is used in research related to obesity and diabetes.
  • Orally active rare sugar alcohol
  • Exhibits anti-obesity activities
  • Exhibits hypoglycaemic activities
  • Reduces carcass and total body fat mass
  • Increases cecal weight, surface area, and short-chain fatty acid production
  • Upregulates cecal microbial enzymes linked to butyrate metabolism
  • Suitable for research on obesity and diabetes
Catalog No. 50-004-18747