Learn More
Medchemexpress LLC HY-117407 10mg Medchemexpress, ALLO-2 CAS:1357350-60-7 Purity:>98%
SDP
Catalog No. 501969472
Encompass_Preferred
Medchemexpress, HY-117407 10mg ALLO-2 CAS:1357350-60-7 ALLO-2 is a potent drug-resistant Smoothened (Smo) mutant antagonist that inhibits Smo agonist Hh-Ag1.5-induced luciferase expression in TM3-Gli-Luc cells with IC50 of 6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9472 Supplier Medchemexpress LLC Supplier No. HY11740710MG
Medchemexpress, HY-117407 10mg ALLO-2 CAS:1357350-60-7 ALLO-2 is a potent drug-resistant Smoothened (Smo) mutant antagonist that inhibits Smo agonist Hh-Ag1.5-induced luciferase expression in TM3-Gli-Luc cells with IC50 of 6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.