Learn More
Medchemexpress LLC ALLO-2 | 1316652-41-1 | 98.7% | 326.37 g·mol⁻¹ | C16H14N4O2S | 25 MG
SDP
Catalog No. 5000226796
Encompass_Preferred
NCC-149 is a selective histone deacetylase 8 (HDAC8) inhibitor used in research on neural differentiation and HDAC8-related signaling pathways. It is supplied as a high-purity solid for laboratory use and is commonly prepared as a DMSO stock for cellular and biochemical assays.
  • Selective HDAC8 inhibition for targeted epigenetic studies.
  • Suitable for neural differentiation experiments and mechanistic research.
  • High reported purity for reproducible experimental results.
  • Available in small milligram pack sizes for screening and assay development.
  • Soluble in DMSO for preparation of concentrated stock solutions.
Catalog No. 50-002-26796 Supplier Medchemexpress LLC Supplier No. HY11734825MG
NCC-149 is a selective histone deacetylase 8 (HDAC8) inhibitor used in research on neural differentiation and HDAC8-related signaling pathways. It is supplied as a high-purity solid for laboratory use and is commonly prepared as a DMSO stock for cellular and biochemical assays.
  • Selective HDAC8 inhibition for targeted epigenetic studies.
  • Suitable for neural differentiation experiments and mechanistic research.
  • High reported purity for reproducible experimental results.
  • Available in small milligram pack sizes for screening and assay development.
  • Soluble in DMSO for preparation of concentrated stock solutions.