Learn More
Medchemexpress LLC HY-117407 5mg Medchemexpress, ALLO-2 CAS:1357350-60-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1174075MG
Encompass_Preferred
Medchemexpress, HY-117407 5mg ALLO-2 CAS:1357350-60-7 ALLO-2 is a potent drug-resistant Smoothened (Smo) mutant antagonist that inhibits Smo agonist Hh-Ag1.5-induced luciferase expression in TM3-Gli-Luc cells with IC 50 of 6 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2520