Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Allocryptopine | 485-91-6 | 99.7% | 369.41 g/mol | C21H23NO5 | 10 MG
SDP

Catalog No. 5000296311
Encompass_Preferred

Allocryptopine is a dibenzazecine alkaloid (C21H23NO5, MW 369.41 g/mol) used as a potassium-channel inhibitor in cardiovascular and ion-channel research. Supplied in small research pack sizes with high reported purity, the compound is intended for in vitro and other laboratory applications as a research reagent.

  • Chemical formula: C21H23NO5
  • Molecular weight: 369.41 g/mol
  • CAS number: 485-91-6
  • Purity: 99.7%
  • Target: Potassium channel inhibitor (hERG-related activity)
  • Intended use: Research use only
  • Typical pack size: 10 MG

Catalog No. 50-002-96311 Supplier Medchemexpress LLC Supplier No. HYN193310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allocryptopine is a dibenzazecine alkaloid (C21H23NO5, MW 369.41 g/mol) used as a potassium-channel inhibitor in cardiovascular and ion-channel research. Supplied in small research pack sizes with high reported purity, the compound is intended for in vitro and other laboratory applications as a research reagent.

  • Chemical formula: C21H23NO5
  • Molecular weight: 369.41 g/mol
  • CAS number: 485-91-6
  • Purity: 99.7%
  • Target: Potassium channel inhibitor (hERG-related activity)
  • Intended use: Research use only
  • Typical pack size: 10 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.